Learn More
MP Biomedicals™ alpha-Amylase from Bacillus subtilis
Liquefying diastase supplied as dry, free-flowing powder with high uniformity and storage stability
99.43 € - 400.68 €
Specifications
Color | Off-White to Tan |
---|---|
Incubator Temperature | 60°C to 80°C |
pH | 5 to 7 |
For Use With (Application) | Carbohydrate hydrolysis |
Content And Storage | 4°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11463130
|
MP Biomedicals™
0210044780 |
100 g |
99.43 €
100g |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11473130
|
MP Biomedicals™
0210044790 |
500 g |
400.68 €
500g |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- CAS Number: 9000-90-2
- EC Number: 232-565-6
- MDL Number: MFCD00081319
- Format: Off-white to tan powder
- Specific Activity: ∽165,000BAU/g
- pH: 5 to 7 (optimum around 6.1)
Specifications
Off-White to Tan | |
5 to 7 | |
4°C | |
alpha-Amylase |
60°C to 80°C | |
Carbohydrate hydrolysis | |
Bacillus subtilis | |
Powder |