missing translation for 'onlineSavingsMsg'
Learn More

Moxifloxacin Hydrochloride, MedChemExpress

Product Code. p-200101171
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16474067

Brand: Medchem Express HY66011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Moxifloxacin Hydrochloride (BAY 12-8039) is an oral 8-methoxyquinolone antimicrobial for use in the treatment of acute bacterial sinusitis, acute bacterial exacerbations of chronic bronchitis, and community-acquired pneumonia.

Moxifloxacin Hydrochloride (BAY 12-8039) is an oral 8-methoxyquinolone antimicrobial for use in the treatment of acute bacterial sinusitis, acute bacterial exacerbations of chronic bronchitis, and community-acquired pneumonia.

Specifications

Chemical Name or Material Moxifloxacin Hydrochloride
Physical Form Solid
CAS 186826-86-8
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C21H25ClFN3O4
For Use With (Application) COVID-19-immunoregulation
Synonym BAY 12-8039
Solubility Information DMSO : ≥ 31 mg/mL (70.79 mM) ∣H2O : 16.67 mg/mL (38.07 mM; Need ultrasonic)
SMILES O=C(C1=CN(C2CC2)C3=C(C=C(F)C(N(C4)C[C@]5([H])[C@@]4([H])CCCN5)=C3OC)C1=O)O.Cl
Molecular Weight (g/mol) 437.89
Formula Weight 437.89
Percent Purity 99.51%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.