Learn More
R&D Systems™ Mouse Wnt-8a (NP_033316.2) VersaClone cDNA
Product Code. 18794207
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18794207 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18794207 Supplier R&D Systems™ Supplier No. RDC3734
  • ONLINE EXCLUSIVE PRICE
    361.00 €
    341.25 € / 10µg
    Save 19.75 €
    5% Off
 Restricted Item
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Gene Wnt-8a
Accession Number NP_033316.2
Gene ID (Entrez) 7478
Gene Alias protein Wnt-8a, Protein Wnt-8d, wingless-type MMTV integration site family, member 8A, Wnt8a, WNT8DWNT8d
Content And Storage Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Product Line R&D Systems
Quantity 10 μg