Learn More
Proteintech Mouse TNFRSF1B ELISA Kit
Product Code. 17264886 Shop All Proteintech Products
Change view
Click to view available options
Gene ID (Entrez):
21938
Unit Size:
96 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gene ID (Entrez) unitSize
17264886 21938 96 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17264886 Supplier Proteintech Supplier No. KE1006196T
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
TNFRSF1B, also known as TNFR2 or CD120b or p75, is a member of the tumor necrosis factor receptor superfamily, which also contains TNFRSF1A.
Tumor necrosis factor (TNF, TNFSF2) is a multifunctional cytokine that plays a key role in regulating inflammation, immune functions, host defense, and apoptosis. TNF signals through TNFRSF1A and TNFRSF1B. TNFRSF1A is widely expressed, whereas TNFRSF1B exhibits more restricted expression, being found on CD4 and CD8 T lymphocytes, endothelial cells, microglia, oligodendrocytes, neuron subtypes, cardiac myocytes, thymocytes and human mesenchymal stem cells. Both TNFRSF1A and TNFRSF1B can also exist in soluble forms, probably derived by proteolytic cleavage from the cell surface forms. Various defects in the TNFRSF1B pathway, due to polymorphisms in the TNFRSF1B gene, upregulated expression of TNFRSF1B and TNFRSF1B shedding, have been implicated in the pathology of several autoimmune disorders.
Specifications
Gene ID (Entrez) 21938
compliance-icons compliance-icons
Product Identifier
  • 96RXN Mouse TNFRSF1B ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine