missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TIMP-1 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
209.00 € - 1935.00 €
Specifications
| Antigen | TIMP-1 |
|---|---|
| Clone | 151620 |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18719597
![]() |
R&D Systems
MAB980-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18796458
![]() |
R&D Systems
MAB980-100 |
100 μg |
487.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18706439
![]() |
R&D Systems
MAB980-500 |
500 μg |
1935.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIMP-1 Monoclonal specifically detects TIMP-1 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| TIMP-1 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMP1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1 | |
| TIMP1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 151620 | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P12032 | |
| 7076 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TIMP-1 Cys25-Arg205 Accession # P12032 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse TIMP-1 in ELISAs. In sandwich ELISAs, no cross-reactivity or interference was observed with recombinant human (rh) TIMP-1, rhTIMP-2, rhTIMP-3, rhTIMP-4, or rrTIMP-1. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title