missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TGF-beta RI/ALK-5 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF587-SP
This item is not returnable.
View return policy
Description
TGF-beta RI/ALK-5 Polyclonal specifically detects TGF-beta RI/ALK-5 in Mouse samples. It is validated for Western Blot.
Specifications
| TGF-beta RI/ALK-5 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AAT5, activin A receptor type II-like kinase, 53kD, activin A receptor type II-like kinase, 53kDa, Activin receptor-like kinase 5, ACVRLK4, ALK-5, ALK-5ALK5, EC 2.7.11, EC 2.7.11.30, LDS1A, LDS2A, Serine/threonine-protein kinase receptor R4, SKR4, TbetaR-I, TGFB1R1, TGF-beta receptor type I, TGF-beta receptor type-1, TGF-beta type I receptor, TGFbetaRI, TGFBR1, TGF-bRI, TGFR-1, transforming growth factor beta receptor I, transforming growth factor, beta receptor 1, transforming growth factor, beta receptor I (activin A receptor type II-likekinase, 53kD), Transforming growth factor-beta receptor type I | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7046 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| BAA05023 | |
| TGFBR1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TGF-beta RI/ALK-5 Ala21-Glu121 Accession # BAA05023 | |
| 25 μg | |
| Primary | |
| Detects mouse TGF-beta RI/ALK-5 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) TGF-beta RI is observed, and less than 2% cross-reactivity with recombinant mouse TGF-beta RII and rhTGF-beta RIII is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction