missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Shrew-1/AJAP1 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF7970-SP
This item is not returnable.
View return policy
Description
Shrew-1/AJAP1 Polyclonal specifically detects Shrew-1/AJAP1 in Mouse samples. It is validated for Western Blot.
Specifications
| Shrew-1/AJAP1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| adherens junction associated protein 1, adherens junction-associated protein 1, adherens junctions associated protein 1, AJAP1, Gm573, membrane protein shrew-1, MOT8, RP3-426F10.1, Shrew1, SHREW-1, transmembrane protein SHREW1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 55966 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| A2ALI5 | |
| AJAP1 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Shrew-1/AJAP1 His157-His283 Accession # A2ALI5 | |
| 25 μg | |
| Primary | |
| Detects mouse Shrew-1/AJAP1 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human AJAP1 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction