missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse SCF/c-kit Ligand Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB455-SP
This item is not returnable.
View return policy
Description
SCF/c-kit Ligand Monoclonal specifically detects SCF/c-kit Ligand in Mouse samples. It is validated for Neutralization.
Specifications
| SCF/c-kit Ligand | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4254 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Neutralization | |
| 40215 | |
| Neutralization 5-30 ug/mL | |
| P20826 | |
| Kitl | |
| E. coli-derived recombinant mouse SCF/c-kit Ligand Lys26-Ala189 Accession # P20826 | |
| 25 μg | |
| Primary | |
| Detects mouse SCF/c-kit Ligand in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human SCF. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction