missing translation for 'onlineSavingsMsg'
Learn More

Mouse SCARA5 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. p-7062444
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18778247 25 μg 25µg
18072642 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18778247 Supplier R&D Systems Supplier No. AF4754SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

SCARA5 Polyclonal specifically detects SCARA5 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCARA5
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8K299
Gene Alias FLJ23907, MGC45780, scavenger receptor class A, member 5 (putative), Tesr, testis expressed scavenger receptor, UNQ2938/PRO28700
Gene Symbols SCARA5
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse SCARA5 Arg83-Pro491 Accession # Q8K299
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286133
Test Specificity Detects mouse SCARA5 in direct ELISAs and Western blots. In direct ELISAs, less than 40% cross-reactivity with recombinant human SCARA5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.