missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Mouse Recombinant G-CSF

Product Code. 17119351
Encompass_Preferred
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
This item is not returnable. View return policy

Product Code. 17119351

Brand: STEMCELL Technologies 78014

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Granulocyte colony-stimulating factor

  • Granulocyte colony-stimulating factor (G-CSF) is a member of the CSF family of glycoproteins that regulate hematopoietic cell proliferation, differentiation, and function
  • It is a key cytokine involved in the production of neutrophils and the stimulation of granulocyte colony formation from hematopoietic progenitor cells
  • G-CSF causes a range of effects including a transient reduction of SDF-1 expression , the activation of metalloproteases that cleave VCAM-1, and the release of norepinephrine from the sympathetic nervous system, leading to the release or mobilization of hematopoietic stem cells from the bone marrow into the periphery
  • The G-CSF receptor is expressed on a variety of hematopoietic cells, including myeloid-committed progenitor cells, neutrophils, granulocytes, and monocytes

Specifications

Name Colony-stimulating factor 3, CSF-3, MGI-1G, Pluripoietin
Quantity 100 μg
Cell Type Cancer Cells and Cell Lines; Hematopoietic Stem and Progenitor Cells; Neurons
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.