missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse LRIG1 (NP_032403) VersaClone cDNA
Description
- Also known as: Img; LIG-1; D6Bwg0781e
- LRIG1 belongs to the LRIG gene family.
- It is widely expressed and appears on the surface of prostatic epithelium, endothelial cells, vascular and visceral smooth muscle, mammary epithelium, cardiac muscle, keratinocytes and neurons.
- LRIG1 is believed to negatively regulate the ErbB family of receptors.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mLrig1 |
| Accession Number | NP_032403 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 3289 bp |
| Gene ID (Entrez) | 26018 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?