missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IL-23 Quantikine ELISA Kit
Click to view available options
Sufficient For:
1 x 96-well Plate
Unit Size:
1 set
Description
The Mouse IL-23 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse IL-23A/IL-23 P19 in cell culture supernates (50 ul), serum (50 ul), heparin plasma (50 ul).
Specifications
Specifications
| Conjugate | HRP |
| Specificity | Natural and Recombinant Mouse IL-23. This assay does not recognize free p19 or p40 subunits. |
| Sufficient For | 1 x 96-well Plate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 83430 |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | IL23, IL-23, IL-23 p19/IL-12 p40, IL-23 A, IL-23-A, IL-23p19, IL-23p19/IL-12p40, IL23P19P19, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-23 subunit alpha, Interleukin-23 subunit p19, JKA3 induced upon T-cell activation, MGC79388, SGRF, SGRFIL-23 subunit alpha |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction