missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-17RB Antibody, R&D Systems™

Rat Monoclonal Antibody
196.00 € - 500.00 €
Specifications
| Antigen | IL-17RB |
|---|---|
| Clone | 152309 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18722658
![]() |
R&D Systems
MAB10401-SP |
25 μg |
196.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18100941
|
R&D Systems
MAB10401 |
500 μg |
500.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-17RB Monoclonal specifically detects IL-17RB in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| IL-17RB | |
| LYOPH | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q9JIP3 | |
| 55540 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-17 RB Arg18-Gly286 (predicted) Accession # Q9JIP3 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse IL-17 RB in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) IL-17 R, recombinant mouse IL-17 R, or rhIL-17 RB is observed. |
| 152309 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CRL4IL-17 receptor homolog 1, cytokine receptor CRL4, Cytokine receptor-like 4, Evi27, EVI27IL-17B receptor, IL-17 RB, IL-17 receptor B, IL-17 RH1, IL17BRinterleukin 17B receptor, IL-17ER, IL17RB, IL-17RB, IL17Rh1, IL-17Rh1, IL17RH1MGC5245, interleukin 17 receptor B, interleukin 17 receptor homolog 1, interleukin-17 receptor B, Interleukin-17B receptor | |
| IL17RB | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title