missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IL-1 beta/IL-1F2 Quantikine HS ELISA Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 96
Description
- A 4.0 hour solid phase ELISA designed to measure mouse IL-1 beta levels in serum and plasma.
- It contains E. coli-expressed recombinant mouse IL-1 beta and antibodies raised against the recombinant protein.
- Results obtained using natural IL-1 beta showed linear curves that were parallel to the standard curves obtained using the Quantikine™ HS kit standards.
- These results indicate that the Quantikine™ HS Immunoassay kit can be used to determine relative mass values for natural mouse IL-1 beta.
- Intra-Assay Precision (Precision within an assay) Three samples of known concentration were tested twenty times on one plate to assess intra-assay precision.
- Inter-Assay Precision (Precision between assays) Three samples of known concentration were tested in twenty separate assays to assess inter-assay precision. Assays were performed by at least three technicians.
Specifications
Specifications
| Assay Sensitivity | 0.312 pg/mL |
| Cross Reactivity | < 0.5% cross-reactivity observed with available related molecules.< 50% cross-species reactivity observed with species tested. |
| Product Type | ELISA |
| Specificity | Natural and recombinant mouse IL-1 beta |
| Gene ID (Entrez) | 3553 |
| Host Species | Mouse |
| Quantity | 1 Kit |
| Sample Volume | Serum (50 μL), EDTA Plasma (50 μL), Heparin Plasma(50 μL) |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction