missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgG1 Isotype Control, APC, Invitrogen™
Mouse Isotype Control
Brand: Invitrogen MG105
Additional Details : Weight : 0.10500kg
Description
Mouse IgG1 Isotype Control for Flow
Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.Specifications
Mouse IgG1 | |
0.1mg/mL | |
PBS with 0.1% sodium azide | |
Mouse | |
500 μL | |
Whole Antibody | |
1 Each | |
Not Applicable | |
Liquid | |
50 Tests |
Flow Cytometry | |
APC | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
purified | |
ASR | |
Primary | |
0.1% Sodium azide | |
2-8°C | |
IgG1 | |
Wet Ice |
Analyte Specific Reagent. Analytical and performance characteristics are not established.