Learn More
Mouse IFN-beta Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB82342-SP
 View more versions of this product
Product Code. 18761817
  • 201.00 € / 25µg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
IFN-beta Monoclonal antibody specifically detects IFN-beta in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFN-beta
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Human embryonic kidney cell HEK293-derived Mouse IFN-beta , Ile22-Asn182, Accession # P01575
25 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG2b
ELISA
944233
ELISA
P01575
Rat
Protein A or G purified from cell culture supernatant
RUO
3456
Mouse
Purified