Learn More
Mouse IFN-beta Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB82342-100
This item is not returnable.
View return policy
Description
IFN-beta Monoclonal antibody specifically detects IFN-beta in Mouse samples. It is validated for ELISA
Specifications
| IFN-beta | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 | |
| Human embryonic kidney cell HEK293-derived Mouse IFN-beta , Ile22-Asn182, Accession # P01575 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 944233 | |
| ELISA | |
| P01575 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3456 | |
| Mouse | |
| Purified |