missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse G-CSFR/CD114 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6039-SP
This item is not returnable.
View return policy
Description
G-CSFR/CD114 Polyclonal specifically detects G-CSFR/CD114 in Mouse samples. It is validated for Western Blot.
Specifications
| G-CSFR/CD114 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD114, CD114 antigen, colony stimulating factor 3 receptor (granulocyte), CSF3R, Csfgr, G-CSF R, G-CSF receptor, GCSFR, GCSFRG-CSF-R, granulocyte colony-stimulating factor receptor | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1441 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| P40223 | |
| Csf3r | |
| Mouse myeloma cell line NS0-derived recombinant mouse GCSFR/CD114 Cys26-Asp626 Accession # P40223 | |
| 25 μg | |
| Primary | |
| Detects mouse G-CSF R/CD114 in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction