missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse G-CSF, AOF Recombinant Protein

Product Code. 17656555
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 17656555

Brand: Invitrogen™ RP87743

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

E. coli Recombinant Protein

G-CSF (Granulocyte colony stimulating factor) is a naturally occurring cytokine that stimulates the production and antibacterial function of neutrophils and monocytes. Human G-CSF is an 18.8 kDa protein containing 175 amino acid residues, and a soluble isoform of the G-CSF receptor has been described. The pleotropic cytokine is produced by activated monocytes, macrophages, endothelial cells, fibroblasts, astrocytes, osteoblasts and bone marrow cells. G-CSF has been shown to have specific effects on the proliferation, differentiation and activation of hematopoietic cells. G-CSF is also expressed by various transformed cells such as carcinoma cells and myeloblastic leukemia cells. G-CSF is encoded by two distinct DNA sequences, resulting in a full size, high activity and a shorter, low activity isoform of G-CSF. G-CSF is highly conserved among species and has been shown to exert its biological functions through interaction with its receptor expressed on the surface of hematopoietic progenitors, neutrophilic granulocytes and certain carcinoma cell lines. Clinical use of G-CSF has been approved for several therapeutic applications, treatment of neonatal infections, therapy of acute myocardial infarction, granulocyte transfusion in patients with neutropenia, in severe infections and sepsis, therapy in chronic autoimmune neutropenia, treatment of acute myeloid leukemias, Sweet's syndrome and AIDS. Further, G-CSF has been shown to be a marker protein for different carcinomas such as bladder cancer and dysfunction of the protein has been linked to Kostmann Syndrome.
  • Purity: >95% by SDS-Page
  • Endotoxin Concentration: AX41 ≤EUs/μg
  • Protein Length: 179 aa.
  • Biological Activity: <50 pg/mL
  • Molecular Weight: 19.1 kDa
  • Animal Origin Free: Yes
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09920
For Use With (Application) Control, ELISA, Gel Shift, Western Blot
Formulation 10 mM sodium citrate with no preservative
Gene ID (Entrez) 12985
Molecular Weight (g/mol) 19.1 kDa
Name Mouse G-CSF, AOF
Quantity 100 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Gene Alias C17orf33; colony stimulating factor 3; colony stimulating factor 3 (granulocyte); CSF 3; CSF3; CSF3OS; Csfg; Filgrastim; GCSF; G-CSF; Granulocyte colony-stimulating factor; granulocyte-colony stimulating factor; gsf3; Lenograstim; MGC45931; MGI-IG; Pluripoietin; RP23-395E10.4
Gene Symbol Csf3
Specific Reactivity Mouse
Conjugate Unconjugated
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.