missing translation for 'onlineSavingsMsg'
Learn More

Mouse EphA7 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18727487
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18727487 25 μg 25µg
18059351 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18727487 Supplier R&D Systems Supplier No. AF608SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

EphA7 Polyclonal specifically detects EphA7 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA7
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q61772
Gene Alias Cek11, Ebk, EC 2.7.10, EC 2.7.10.1, EHK3, EHK-3, EK11, EPH homology kinase 3, Eph homology kinase-3, EPH receptor A7, EphA7, EPH-like kinase 11, ephrin type-A receptor 7, HEK11, Mdk1, receptor protein-tyrosine kinase HEK11, tyrosine-protein kinase receptor EHK-3
Gene Symbols EPHA7
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse EphA7 isoform 1 Ala30-Pro554 (Gly540-Glu544 del) Accession # Q61772
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2045
Test Specificity Detects mouse EphA7 in direct ELISAs and Western blots. In direct ELISAs, approximately 2% cross-reactivity with recombinant mouse (rm) EphA3 is observed and less than 1% cross-reactivity with rmEphA2, rmEphA4, rmEphA6, and rmEphA8 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.