missing translation for 'onlineSavingsMsg'
Learn More

Mouse DPPA5/ESG1 Antibody, R&D Systems™

Product Code. p-7062038
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18748527 25 μg 25µg
18030852 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18748527 Supplier R&D Systems Supplier No. AF3984SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

DPPA5/ESG1 Polyclonal specifically detects DPPA5/ESG1 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPPA5/ESG1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9CQS7
Gene Alias developmental pluripotency associated 5, developmental pluripotency-associated 5 protein, embryonal stem cell specific gene 1, Embryonal stem cell-specific gene 1 protein, Esg1, ESG1ESG-1, hDPPA5
Gene Symbols Dppa5a
Host Species Goat
Immunogen E. coli-derived recombinant mouse DPPA5/ESG1 Met1-Glu118 Accession # Q9CQS7
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340168
Test Specificity Detects mouse DPPA5/ESG1 in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human DPPA5 is observed and approximately 10% cross-reactivity with recombinant mouse DPPA4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.