Learn More
R&D Systems™ Mouse Dopamine D2 R/DRD2 (NP_034207.2) VersaClone cDNA
Product Code. 18741077
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18741077 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18741077 Supplier R&D Systems™ Supplier No. RDC3500
Please to purchase this item. Need a web account? Register with us today!
Specifications
Gene Dopamine D2R/DRD2
Accession Number NP_034207.2
Gene ID (Entrez) 1813
Gene Alias D(2) dopamine receptor, D2DR, D2R, Dopamine D2 receptor, Dopamine D2R, dopamine receptor D2, dopamine receptor D2 isoform, DRD2, seven transmembrane helix receptor
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg