missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL2/MIP-2 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 16 publications
Brand: R&D Systems AF-452-NA
This item is not returnable.
View return policy
Description
CXCL2/GRO beta/MIP-2/CINC-3 Polyclonal specifically detects CXCL2/GRO beta/MIP-2/CINC-3 in Mouse samples. It is validated for Immunohistochemistry, Neutralization, Immunocytochemistry.
Specifications
| CXCL2/GRO beta/MIP-2/CINC-3 | |
| Polyclonal | |
| Unconjugated | |
| P10889 | |
| CXCL2 | |
| E. coli-derived recombinant mouse CXCL2/MIP-2 Ala28-Asn100 Accession # P10889 | |
| 100 μg | |
| Primary | |
| Detects mouse CXCL2/MIP-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) GRO alpha , rhGRO beta , rhGRO gamma , recombinant rat (rr) CINC-1, rrCINC-2 alpha , rrCINC-2 beta , and rrCINC-3 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.015-0.075 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| chemokine (C-X-C motif) ligand 2, CINC-2a, CINC3, CINC-3, C-X-C Motif Chemokine 2, GRO beta, GRO2, GRO2 oncogene, GROb, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MGSA-beta, MIP2, MIP-2, MIP-2a, MIP2A, MIP2-alpha, SCYB2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2920 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction