missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Crossveinless-2/CV-2 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF2299
This item is not returnable.
View return policy
Description
Crossveinless-2/CV-2/BMPER Polyclonal specifically detects Crossveinless-2/CV-2/BMPER in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Crossveinless-2/CV-2/BMPER | |
| Polyclonal | |
| Unconjugated | |
| AAH66153 | |
| BMPER | |
| Mouse myeloma cell line NS0-derived recombinant mouse Crossveinless-2/CV-2 (R&D Systems, Catalog # 2299-CV) Ala39-Arg685 Accession # AAH66153 | |
| 100 μg | |
| Primary | |
| Detects mouse Crossveinless-2/CV-2 in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human CV-2 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| BMP binding endothelial regulator, BMP-binding endothelial regulator precursor protein, BMP-binding endothelial regulator protein, BMPER, Bone morphogenetic protein-binding endothelial cell precursor-derived regulator, CRIM3, crossveinless 2, crossveinless-2, Cv2, CV-2, hCV2, KIAA1965, Protein crossveinless-2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 168667 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction