Learn More
R&D Systems™ Mouse CL-L1/COLEC10 (NP_775598.2) VersaClone cDNA
Specifications
Specifications
| Gene | COLEC10 |
| Accession Number | NP_775598.2 |
| Gene ID (Entrez) | 10584 |
| Gene Alias | CL-34, CL-L1, CLL1collectin 34, COLEC10, collectin liver 1, Collectin liver protein 1, collectin sub-family member 10 (C-type lectin), collectin-10, Collectin-34, MGC118794, MGC118795 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |