missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cardiotrophin-1/CT-1 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB438
This item is not returnable.
View return policy
Description
Cardiotrophin-1/CT-1 Monoclonal specifically detects Cardiotrophin-1/CT-1 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| Cardiotrophin-1/CT-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cardiophin 1, cardiotrophin 1, cardiotrophin-1, Cardiotrophin1, CT-1CT1, CTF1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1489 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, ELISA | |
| 91907 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q60753 | |
| CTF1 | |
| E. coli-derived recombinant mouse Cardiotrophin-1 Ser2-Ala203 Accession # Q60753 | |
| 500 μg | |
| Primary | |
| Detects mouse Cardiotrophin-1 in ELISAs and Western blots. In ELISAs, this antibody shows approximately 20% cross-reactivity with recombinant human (rh) Cardiotrophin-1 and no cross-reactivity with rmIL-11, rmIL-6, rrCNTF, rmLIF, or rmOSM. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction