missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitellogenin (VTG), Mouse, Clone: 1G2, Abnova™
Mouse monoclonal antibody raised against native vitellogenin (VTG).
Brand: Abnova MAB1727.100ug
This item is not returnable.
View return policy
Specifications
| Vitellogenin (VTG) | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) ELISA (0.1-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Liquid |
| ELISA, Western Blot | |
| 1G2 | |
| Mouse monoclonal antibody raised against native vitellogenin (VTG). | |
| In PBS (0.1% BSA, 0.1% proclin) | |
| Native from carp lipovitellin from eggs. | |
| RUO | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction