missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VCAM-1/CD106 Mouse anti-Human, Clone: HAE-2Z, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB809-500
This item is not returnable.
View return policy
Description
VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| VCAM-1/CD106 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| VCAM1 | |
| Human VCAM-1 | |
| 500 μg | |
| Primary | |
| Detects recombinant human VCAM-1 in direct ELISAs and Western blots. | |
| Human | |
| Supernatant |
| ELISA | |
| HAE-2Z | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7412 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?