missing translation for 'onlineSavingsMsg'
Learn More

UBTF, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16187225
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16187225 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16187225 Supplier Abnova Supplier No. H00007343A01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a partial recombinant UBTF.

Upstream binding factor (UBF) is a transcription factor required for expression of the 18S, 5.8S, and 28S ribosomal RNAs, along with SL1 (a complex of TBP (MIM 600075) and multiple TBP-associated factors or 'TAFs'). Two UBF polypeptides, of 94 and 97kDa, exist in the human (Bell et al., 1988 [PubMed 3413483]). UBF is a nucleolar phosphoprotein with both DNA binding and transactivation domains. Sequence-specific DNA binding to the core and upstream control elements of the human rRNA promoter is mediated through several HMG boxes (Jantzen et al., 1990 [PubMed 2330041]).[supplied by OMIM

Sequence: PPAATNSSKKMKFQGEPKKPPMNGYQKFSQELLSNGELNHLPLKERMVEIGSRWQRISQSQKEHYKKLAEEQQKQYKVHLDLWVKSLSPQDRAAYKEYIS

Specifications

Antigen UBTF
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant UBTF.
Formulation 50% glycerol
Gene UBTF
Gene Accession No. NM_014233
Gene Alias NOR-90/UBF
Gene Symbols UBTF
Host Species Mouse
Immunogen UBTF (NP_055048, 551 a.a. ∽ 650 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7343
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.