Learn More
SPN, Mouse, Clone: TP1/36, Abnova™
Mouse monoclonal antibody raised against native SPN.
Brand: Abnova MAB4980.100ug
Description
Sialophorin (leukosialin) is a major sialoglycoprotein on the surface of human T lymphocytes, monocytes, granulocytes, and some B lymphocytes, which appears to be important for immune function and may be part of a physiologic ligand-receptor complex involved in T-cell activation.[supplied by OMIM
Specifications
| SPN | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user. | |
| SPN | |
| SPN | |
| Native purified SPN from myeloblastic cell line KG1. | |
| RUO | |
| 6693 | |
| Store in the dark at 4°C. Avoid prolonged exposure to light. | |
| IgG1 |
| Flow Cytometry, Immunofluorescence | |
| TP1/36 | |
| Mouse monoclonal antibody raised against native SPN. | |
| In buffer containing 1% BSA, pH 7.2 (0.09% sodium azide) | |
| CD43/GPL115/LSN | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.