missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat IgG1 Heavy Chain Secondary Antibody (MARG1-2), Janelia Fluor 549, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG1 Heavy Chain Monoclonal antibody specifically detects IgG1 Heavy Chain in Rat samples. It is validated for ELISA
Specifications
Specifications
| Antigen | IgG1 Heavy Chain |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | MARG1-2 |
| Conjugate | Janelia Fluor 549 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Rat IR27 IgG1 myeloma protein |
| Purification Method | Affinity purified |
| Quantity | 0.25 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?