missing translation for 'onlineSavingsMsg'
Learn More

PTGIS, Mouse, Monoclonal Antibody, Abnova™

Product Code. 16065996
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16065996 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16065996 Supplier Abnova Supplier No. MAB3798.20ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against PTGIS.

Specifications

Antigen PTGIS
Applications ELISA, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against PTGIS.
Dilution ELISA (1:10) Immunoprecipitation (1:2) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Gene PTGIS
Gene Alias MGC140775
Gene Symbols PTGIS
Host Species Mouse
Immunogen Bovine PTGIS.
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282021
Test Specificity Bovine PGI2 Synthase reacts to most mammalian species, including bovine, mouse, rat, sheep, guinea pig, and rabbit.
Target Species Bovine, Guinea Pig, Mouse, Rabbit, Rat, Sheep
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.