Learn More
PAX7, Mouse, Clone: 3F10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PAX7.
Brand: Abnova H00005081-M14A.200uL
Additional Details : Weight : 0.02000kg
Description
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 7 gene is unknown but speculated to involve tumor suppression since fusion of this gene with a forkhead domain family member has been associated with alveolar rhabdomyosarcoma. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: AYGARHSFSSYSDSFMNPAAPSNHMNPVSNGLSPQVMSILGNPSAVPPQPQADFSISPLHGGLDSATSISASCSQRADSIKPGDSpecifications
PAX7 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_002584 | |
PAX7 | |
PAX7 (NP_001128726.1,351 a.a. ∽ 434 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
5081 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
3F10 | |
Mouse monoclonal antibody raised against a partial recombinant PAX7. | |
PAX7 | |
HUP1/PAX7B | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |