missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF200, Mouse, Clone: NF-200, Abnova™
Mouse monoclonal antibody raised against native NF200.
Brand: Abnova MAB1523.100ug
This item is not returnable.
View return policy
Specifications
| NF200 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Immunohistochemistry (Frozen sections) (1-2 ug/mL) Western Blot (0.5 mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| Affinity purification | |
| RUO | |
| Human, Mouse, Pig, Rat | |
| Lyophilized |
| Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot | |
| N52 | |
| Mouse monoclonal antibody raised against native Nefh. | |
| Lyophilized from 1.2% sodium acetate (2mg BSA, 0.01mg sodium azide) | |
| Native purified pig neurofilament 200. | |
| 100 μg | |
| Primary | |
| Store at -20°C on dry atmosphere. After reconstitution with 1mL of 1.2% sodium acetate or neutral PBS to a concentration of 100μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction