missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlamydia trachomatis MOMP, Mouse, Clone: CL12-755.1.31.3, Abnova™
Mouse monoclonal antibody raised against Chlamydia trachomatis MOMP.
Brand: Abnova MAB6160.100ug
This item is not returnable.
View return policy
Specifications
| Chlamydia trachomatis MOMP | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Bacteria | |
| Liquid |
| ELISA, Immunofluorescence | |
| CL12-755.1.31.3 | |
| Mouse monoclonal antibody raised against Chlamydia trachomatis MOMP. | |
| In PBS, pH 7.4 | |
| Chlamydia trachomatis elementary bodies. | |
| RUO | |
| These antibodies recognize Chlamydia trachomatis serovars A, B, Ba, C, D, E, F, G, H, I, J, K, L1, L2, L3. | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction