missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYKK, Mouse, Clone: 4F2, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant HYKK.
Brand: Abnova H00123688-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MSSGNYQQSEALSKPTFSEEQASALVESVFGLKVSKVRPLPSYDDQNFHVYVSKTKDGPTEYVLKISNTKASKNPDLIEVQNHIIMFLKAAGFPTASVCHTKGDNTASLVSVDSGSEIKSYLVRLLTYLPGRPIAELPVSPQLLYEIGKLAAKLDKTLQEGKPRVTPLLAKNSpecifications
| HYKK | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| ENST00000360519 | |
| HYKK | |
| HYKK (ENSP00000353710,1 a.a. ∽ 172 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 4F2 | |
| Mouse monoclonal antibody raised against a full-length recombinant HYKK. | |
| HYKK | |
| AGPHD1 | |
| Mouse | |
| Protein A Purification | |
| RUO | |
| 123688 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction