missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG3 Heavy Chain Secondary Antibody (5G12cc), PE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG3 Heavy Chain Monoclonal antibody specifically detects IgG3 Heavy Chain in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | IgG3 Heavy Chain |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 5G12cc |
| Conjugate | PE |
| Formulation | PBS |
| Host Species | Mouse |
| Immunogen | Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunised with IgG3 from human serum. |
| Purification Method | Protein A purified |
| Quantity | 0.2 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?