missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP, Mouse, Clone: F56-BA1.2.3, Abnova™
Mouse monoclonal antibody raised against full-length recombinant GFP.
Brand: Abnova MAB1885.100ug
This item is not returnable.
View return policy
Specifications
| GFP | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Liquid |
| Western Blot | |
| F56-BA1.2.3 | |
| Mouse monoclonal antibody raised against full length recombinant GFP. | |
| In PBS (0.08% sodium azide) | |
| Recombinant protein corresponding to full length GFP. | |
| RUO | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction