missing translation for 'onlineSavingsMsg'
Learn More

A1/A2, Mouse, Clone: Z2A, Abnova™

Product Code. 16260950
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16260950 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16260950 Supplier Abnova Supplier No. MAB5411.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against native A1/A2.

Specifications

Antigen A1/A2
Applications Agglutination, ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Z2A
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native A1/A2.
Dilution Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 300mM NaCl, 1.25mM EDTA (0.09% sodium azide)
Host Species Mouse
Immunogen Native human blood A1/A2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity A1, A2 human blood antigen.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.