missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Angiopoietin-like Protein 3/ANGPTL3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF136-SP
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 3/ANGPTL3 Polyclonal specifically detects Angiopoietin-like Protein 3/ANGPTL3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Angiopoietin-like Protein 3/ANGPTL3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AGNPT5, Ang-5, angiopoietin 5, Angiopoietin-5, angiopoietin-like 3, Angiopoietin-like protein 3, angiopoietin-related protein 3, ANGPT5angiopoietin-5, ANGPTL3, FHBL2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27329 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q9R182 | |
| ANGPTL3 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Angiopoietin-like Protein 3/ANGPTL3 Ser17-Thr455 Accession # Q9R182 | |
| 25 μg | |
| Primary | |
| Detects mouse Angiopoietin-like Protein 3/ANGPTL3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Angiopoietin-1, rhAngiopoietin-2, recombinant mouse Angiopoietin-3, rhAngiopoietin-4, and rhAngiopoietin-like factor/CDT6 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction