Learn More
R&D Systems™ Mouse alpha 1B-Glycoprotein (NP_001074536) VersaClone cDNA
Product Code. 18796739
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18796739 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18796739 Supplier R&D Systems™ Supplier No. RDC2011
Please to purchase this item. Need a web account? Register with us today!
R&D Systems™ Mouse alpha 1B-Glycoprotein (NP_001074536) VersaClone cDNA is a plasma glycoprotein of unknown function.
The Mouse alpha 1B-Glycoprotein (NP_001074536) VersaClone cDNA clone contains the complete ORF for alpha 1B-Glycoprotein, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The alpha 1B-Glycoprotein insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY
Specifications
Gene mA1bg
Accession Number NP_001074536
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl
Insert Type 1552bp
pH 8.5
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg