missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Mouse Alkaline Phosphatase, Liver/Bone/Kidney ELISA Kit (Chemiluminescence)

Product Code. 18109312 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18109312

Brand: Novus Biologicals™ NBP269933

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Mouse Alkaline Phosphatase, Liver/Bone/Kidney ELISA Kit (Chemiluminescence) measures mouse Alkaline Phosphatase, Liver/Bone/Kidney in serum, plasma and other biological fluids.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.25 to 2000 pg/mL
Assay Sensitivity 18.75 pg/mL
Cross Reactivity This kit recognizes Mouse BALP in samples. No significant cross-reactivity or interference between Mouse BALP and analogues was observed.
Product Type ELISA Kit (Chemiluminescence)
Sample Type Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 249
Gene Symbol ALPL
Host Species Mouse
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Alkaline Phosphatase, Liver/Bone/Kidney
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.