missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Adiponectin/Acrp30 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB1119-SP
This item is not returnable.
View return policy
Description
Adiponectin/Acrp30 Monoclonal specifically detects Adiponectin/Acrp30 in Mouse samples. It is validated for Western Blot.
Specifications
| Adiponectin/Acrp30 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q60994 | |
| ADIPOQ | |
| Mouse myeloma cell line NS0-derived recombinant mouse Adiponectin Glu18-Asn247 Accession # Q60994 | |
| 25 μg | |
| Primary | |
| Detects full-length mouse Adiponectin (aa 18-247) in direct ELISAs and Western blots. Shows no cross-reactivity with recombinant human Adiponectin. Does not detect the C-terminal globular domain of mouse Adiponectin (aa 111-247). | |
| Mouse | |
| Purified |
| Western Blot | |
| 173205 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACDC, Acrp30, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, ApM1, APM1APM-1, C1q and collagen domain-containing protein, GBP28, GBP28apM1, Gelatin-binding protein | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9370 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction