Learn More
MedChemExpress Morusin
Click to view available options
:
2 mg
Specifications
Specifications
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| Molecular Weight (g/mol) | 420.45 g/mol |
| Shipping Condition | Room temperature |
| For Use With (Application) | Cancer-Kinase/protease |
| CAS | 62596-29-6 |
| Product Type | Natural Products |
| Purity | 99.83% |
| Solubility | DMSO : ≥ 100 mg/mL (237.84 mM) |
| Synonym | Mulberrochromene |
| Target | NF-κB∣STAT∣Bacterial |
| Show More |