Learn More
MedChemExpress Morusin
Product Code. 16419048
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16419048 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16419048 Supplier MedChemExpress Supplier No. HYN0622
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress Morusin is a prenylated flavonoid isolated from M. australis with various biological activities, such as antitumor, antioxidant, and anti-bacteria property. Morusin could inhibit NF-κB and STAT3 activity.
Specifications
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 420.45 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 62596-29-6
Product Type Natural Products
Purity 99.83%
Solubility DMSO : ≥ 100 mg/mL (237.84 mM)
Synonym Mulberrochromene
Target NF-κB∣STAT∣Bacterial
Quantity 10 mg