missing translation for 'onlineSavingsMsg'
Learn More

Morinidazole, MedChemExpress

Product Code. 16416838
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16416838

Brand: Medchem Express HY15781

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Morinidazole is a novel 5-nitroimidazole antimicrobial drug that undergoes extensive metabolism in humans via N+-glucuronidation and sulfation, for the treatment of bacterial infections including appendicitis and pelvic inflammatory disease (PID) caused by anaerobic bacteria.

Morinidazole is a novel 5-nitroimidazole antimicrobial drug that undergoes extensive metabolism in humans via N+-glucuronidation and sulfation, for the treatment of bacterial infections including appendicitis and pelvic inflammatory disease (PID) caused by anaerobic bacteria.

Specifications

Chemical Name or Material Morinidazole
Physical Form Solid
CAS 92478-27-8
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C11H18N4O4
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (369.97 mM)
SMILES OC(CN1C([N+]([O-])=O)=CN=C1C)CN2CCOCC2
Molecular Weight (g/mol) 270.29
Formula Weight 270.29
Percent Purity 98.05%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.