missing translation for 'onlineSavingsMsg'
Learn More

mOrange/mOrange2 Mouse anti-Tag, Clone: OTI4E10 (formerly 4E10), TrueMAB™, Origene

Product Code. 17610201
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17610201 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17610201 Supplier Origene Technologies Supplier No. CF180053

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

mOrange/mOrange2 Monoclonal antibody specifically detects mOrange/mOrange2 in Tag samples. It is validated for Western Blot

Specifications

Antigen mOrange/mOrange2
Applications Western Blot
Classification Monoclonal
Clone OTI4E10 (formerly 4E10)
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative
Gene Alias mOrange
Host Species Mouse
Immunogen This antibody is raised against a recombinant protein of mOrange expressed in HEK293 cells. It recognizes both mOrange and mOrange2 proteins.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.