Learn More
Monoethyl Ester 1,2-Ethanedisulfonic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Monoethyl Ester 1,2-Ethanedisulfonic Acid |
| CAS | 857728-63-3 |
| Molecular Formula | C4 H10 O6 S2 |
| Quantity | 250 mg |
| Synonym | 1-Ethyl Ester 1,2-Ethanedisulfonic Acid |
| InChI Formula | InChI=1S/C4H10O6S2/c1-2-10-12(8,9)4-3-11(5,6)7/h2-4H2,1H3,(H,5,6,7) |
| SMILES | CCOS(=O)(=O)CCS(=O)(=O)O |
| IUPAC Name | 2-ethoxysulfonylethanesulfonic acid |
| Molecular Weight (g/mol) | 218.25 |
| Formula Weight | 217.9919 |