Learn More
Monodecyl Phthalate-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Monodecyl Phthalate-d4 |
| CAS | 2732762-53-5 |
| Molecular Formula | C18 D4 H22 O4 |
| Quantity | 5 mg |
| Synonym | Monodecyl Phthalate-d4, 1-Decyl 1,2-benzenedicarboxylate-3,4,5,6-d4, Phthalic-d4 acid monodecyl ester, 2-(Decyloxycarbonyl)benzoic-3,4,5,6-d4 acid, Decyl hydrogen phthalate, Decyl phthalate-d4, Capryl phthalate-d4, Monodecyl Phthalate-3,4,5,6-D4, mono-n-Decyl Phthalate-3,4,5,6-d4 |
| InChI Formula | InChI=1S/C18H26O4/c1-2-3-4-5-6-7-8-11-14-22-18(21)16-13-10-9-12-15(16)17(19)20/h9-10,12-13H,2-8,11,14H2,1H3,(H,19,20)/i9D,10D,12D,13D |
| SMILES | [2H]c1c([2H])c([2H])c(C(=O)OCCCCCCCCCC)c(C(=O)O)c1[2H] |
| IUPAC Name | 2-decoxycarbonyl-3,4,5,6-tetradeuteriobenzoic acid |
| Molecular Weight (g/mol) | 310.4213 |
| Formula Weight | 310.2082 |