Learn More
Monodecarboxy Piperacilloic Acid (Mixture of Diastereomers), TRC
Product Code. 30468565
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30468565 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30468565 Supplier Toronto Research Chemicals Supplier No. TRCM52670050MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Monodecarboxy Piperacilloic Acid (Mixture of Diastereomers)
CAS 64817-23-8
Molecular Formula C22 H29 N5 O6 S
Quantity 50 mg
Synonym α-(4-Ethyl-2,3-dioxo-1-piperazinecarboxamido)benzylpenilloic Acid, [2R-[2α(R*),4β]]-2-[[[[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]phenylacetyl]amino]methyl]-5,5-dimethyl-4-thiazolidinecarboxylic Acid, Piperacillin EP Impurity C
InChI Formula InChI=1S/C22H29N5O6S/c1-4-26-10-11-27(19(30)18(26)29)21(33)25-15(13-8-6-5-7-9-13)17(28)23-12-14-24-16(20(31)32)22(2,3)34-14/h5-9,14-16,24H,4,10-12H2,1-3H3,(H,23,28)(H,25,33)(H,31,32)/t14-,15-,16+/m1/s1
SMILES CCN1CCN(C(=O)N[C@@H](C(=O)NC[C@@H]2N[C@@H](C(=O)O)C(C)(C)S2)c3ccccc3)C(=O)C1=O
IUPAC Name (2R,4S)-2-[[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetyl]amino]methyl]-5,5-dimethyl-1,3-thiazolidine-4-carboxylic acid
Molecular Weight (g/mol) 491.561
Formula Weight 491.184